Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGACCCTTAAACAAATATGTCTA[C/T]GTTCACTGTCCAGAATATAATATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
65 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615406 MIM: 611514 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG12-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GNG12-AS1 - GNG12 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WLS - wntless Wnt ligand secretion mediator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002292.3 | Intron | NP_001002292.3 | ||||
NM_001193334.1 | Intron | NP_001180263.1 | ||||
NM_024911.6 | Intron | NP_079187.3 | ||||
XM_011542191.2 | Intron | XP_011540493.1 | ||||
XM_011542192.2 | Intron | XP_011540494.1 | ||||
XM_017002390.1 | Intron | XP_016857879.1 |