Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGCTGGGGAAGGTAATGTTACC[C/T]CTGCAAACAGATGCTCTGGTAAGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615405 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GNG12 - G protein subunit gamma 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018841.5 | 1489 | UTR 3 | NP_061329.3 | |||
XM_017001809.1 | 1489 | UTR 3 | XP_016857298.1 | |||
XM_017001810.1 | 1489 | UTR 3 | XP_016857299.1 | |||
XM_017001811.1 | 1489 | UTR 3 | XP_016857300.1 |
Set Membership: |
HapMap |