Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCCCGCTGCCCGCTGAGCCCCT[A/C]CTGGCCTTGGCCGGGCCGGCAGCCG
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 601094 | ||||||||||||||||||||||||||
Literature Links: |
FOXE3 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
FOXE3 - forkhead box E3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012186.2 | 1102 | Silent Mutation | CTA,CTC | L,L 286 | NP_036318.1 |
LINC01389 - long intergenic non-protein coding RNA 1389 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |