Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTCTTATGAAACCCACTCTGCC[C/T]ACAGTAGTTTATCTGCACATCTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603335 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNAH5 - dynein axonemal heavy chain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001369.2 | Intron | NP_001360.1 | ||||
XM_005248262.3 | Intron | XP_005248319.2 | ||||
XM_017009177.1 | Intron | XP_016864666.1 | ||||
XM_017009178.1 | Intron | XP_016864667.1 | ||||
XM_017009179.1 | Intron | XP_016864668.1 | ||||
XM_017009180.1 | Intron | XP_016864669.1 | ||||
XM_017009181.1 | Intron | XP_016864670.1 | ||||
XM_017009182.1 | Intron | XP_016864671.1 | ||||
XM_017009183.1 | Intron | XP_016864672.1 | ||||
XM_017009184.1 | Intron | XP_016864673.1 | ||||
XM_017009185.1 | Intron | XP_016864674.1 | ||||
XM_017009186.1 | Intron | XP_016864675.1 | ||||
XM_017009187.1 | Intron | XP_016864676.1 | ||||
XM_017009188.1 | Intron | XP_016864677.1 |