Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCATGTGATAGAAAGAGCTGAC[A/G]TAAGAGAAAAGACCCCCACACAGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602949 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAP18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SAP18 - Sin3A associated protein 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SKA3 - spindle and kinetochore associated complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166017.1 | 2017 | UTR 3 | NP_001159489.1 | |||
NM_145061.5 | 2017 | UTR 3 | NP_659498.4 | |||
XM_005266288.3 | 2017 | UTR 3 | XP_005266345.1 | |||
XM_011534994.1 | 2017 | UTR 3 | XP_011533296.1 |