Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAAAAAAAGATTGGTAAACAAA[C/T]GTGAATAGACAAATGTAAAATCAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604242 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RNF6 - ring finger protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005977.3 | Intron | NP_005968.1 | ||||
NM_183043.2 | Intron | NP_898864.1 | ||||
NM_183044.2 | Intron | NP_898865.1 | ||||
NM_183045.1 | Intron | NP_898866.1 | ||||
XM_005266485.2 | Intron | XP_005266542.1 | ||||
XM_005266486.2 | Intron | XP_005266543.1 | ||||
XM_011535177.2 | Intron | XP_011533479.1 | ||||
XM_011535178.1 | Intron | XP_011533480.1 | ||||
XM_017020685.1 | Intron | XP_016876174.1 | ||||
XM_017020686.1 | Intron | XP_016876175.1 | ||||
XM_017020687.1 | Intron | XP_016876176.1 |