Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAN3 - PAN3 poly(A) specific ribonuclease subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175854.7 | Intron | NP_787050.6 | ||||
XM_005266332.3 | Intron | XP_005266389.1 | ||||
XM_005266333.3 | Intron | XP_005266390.1 | ||||
XM_005266334.1 | Intron | XP_005266391.1 | ||||
XM_011535032.2 | Intron | XP_011533334.1 | ||||
XM_011535033.2 | Intron | XP_011533335.1 | ||||
XM_011535034.2 | Intron | XP_011533336.1 | ||||
XM_017020528.1 | Intron | XP_016876017.1 | ||||
XM_017020529.1 | Intron | XP_016876018.1 | ||||
XM_017020530.1 | Intron | XP_016876019.1 | ||||
XM_017020531.1 | Intron | XP_016876020.1 | ||||
XM_017020532.1 | Intron | XP_016876021.1 | ||||
XM_017020533.1 | Intron | XP_016876022.1 |
Set Membership: |
HapMap |