Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCAATATGGCAGGAGAACAAGT[C/T]TGACTTTTTTCCGCAACTAGTGTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605766 MIM: 605661 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLEU2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DLEU2 - deleted in lymphocytic leukemia 2 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3613 - microRNA 3613 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM13 - tripartite motif containing 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007278.2 | Intron | NP_001007279.1 | ||||
NM_005798.4 | Intron | NP_005789.2 | ||||
NM_052811.3 | Intron | NP_434698.1 | ||||
NM_213590.2 | Intron | NP_998755.1 |
Set Membership: |
HapMap |