Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCATTCTGTAGCTTGAATTTTGT[C/T]ATTAGCAAGAAATATTGTTAGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607865 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SETDB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SETDB2 - SET domain bifurcated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160308.2 | Intron | NP_001153780.1 | ||||
NM_001320699.1 | Intron | NP_001307628.1 | ||||
NM_031915.2 | Intron | NP_114121.2 |
SETDB2-PHF11 - SETDB2-PHF11 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320727.1 | Intron | NP_001307656.1 |
Set Membership: |
HapMap |