Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAACTTAATGGAAAGAAGCCCC[G/T]GCAATTATATGCAAAACTGTATATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606013 MIM: 613542 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXO5 - F-box protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTRF1L - mitochondrial translational release factor 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114184.2 | Intron | NP_001107656.1 | ||||
NM_001301047.2 | Intron | NP_001287976.1 | ||||
NM_001301870.1 | Intron | NP_001288799.1 | ||||
NM_001301871.1 | Intron | NP_001288800.1 | ||||
NM_001301872.1 | Intron | NP_001288801.1 | ||||
NM_019041.6 | Intron | NP_061914.3 | ||||
XM_017010959.1 | Intron | XP_016866448.1 | ||||
XM_017010960.1 | Intron | XP_016866449.1 |
Set Membership: |
HapMap |