Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCATTTTCAAATTGTTTTGCTTT[C/G]TAGTTTAAATTCATAAGAGAAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616805 MIM: 192320 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYCT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYCT1 - myc target 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VIP - vasoactive intestinal peptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003381.3 | 1175 | Intron | NP_003372.1 | |||
NM_194435.2 | 1175 | Intron | NP_919416.1 | |||
XM_005267135.3 | 1175 | Intron | XP_005267192.1 | |||
XM_006715562.3 | 1175 | UTR 5 | XP_006715625.1 |
Set Membership: |
HapMap |