Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACGGAGCTGCGGAGGAGCGGGTC[C/T]CGGGATGTGACCGGGGCTCTGCTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615353 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC28A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC28A - coiled-coil domain containing 28A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015439.2 | 281 | Silent Mutation | TCC,TCT | S,S 42 | NP_056254.1 | |
XM_011535728.2 | 281 | Silent Mutation | TCC,TCT | S,S 42 | XP_011534030.1 |
GVQW2 - GVQW motif containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242740.1 | 281 | Intron | NP_001229669.1 |
Set Membership: |
HapMap |