Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGCCCCCGGGCTGGAGTAAGA[G/T]CCGAGCACCGGCGCAGCCTGCGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602532 | ||||||||||||||||||||
Literature Links: |
RNF217 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF217 - ring finger protein 217 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286398.2 | 377 | Missense Mutation | AGC,ATC | S,I 81 | NP_001273327.1 | |
NM_152553.4 | 377 | Intron | NP_689766.1 | |||
XM_011535494.2 | 377 | Missense Mutation | AGC,ATC | S,I 81 | XP_011533796.1 | |
XM_011535495.2 | 377 | Missense Mutation | AGC,ATC | S,I 81 | XP_011533797.1 | |
XM_011535496.2 | 377 | Missense Mutation | AGC,ATC | S,I 81 | XP_011533798.1 | |
XM_011535499.2 | 377 | Missense Mutation | AGC,ATC | S,I 81 | XP_011533801.1 | |
XM_011535500.2 | 377 | Intron | XP_011533802.1 | |||
XM_017010317.1 | 377 | Missense Mutation | AGC,ATC | S,I 81 | XP_016865806.1 |
RNF217-AS1 - RNF217 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |