Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGGTAATATATTTTCTGTGGCA[A/G]AATTTTTTGATTTCTGATCTATGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600970 | ||||||||||||||||||||
Literature Links: |
MYO6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYO6 - myosin VI | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300899.1 | Intron | NP_001287828.1 | ||||
NM_004999.3 | Intron | NP_004990.3 | ||||
XM_005248719.3 | Intron | XP_005248776.1 | ||||
XM_005248720.3 | Intron | XP_005248777.1 | ||||
XM_005248721.3 | Intron | XP_005248778.1 | ||||
XM_005248722.3 | Intron | XP_005248779.1 | ||||
XM_005248724.3 | Intron | XP_005248781.1 | ||||
XM_005248726.3 | Intron | XP_005248783.1 | ||||
XM_017010899.1 | Intron | XP_016866388.1 |