Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607408 MIM: 607415 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAOA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DAOA - D-amino acid oxidase activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161812.1 | 303 | UTR 5 | NP_001155284.1 | |||
NM_001161814.1 | 303 | UTR 5 | NP_001155286.1 | |||
NM_172370.4 | 303 | Missense Mutation | AAG,GAG | K,E 62 | NP_758958.3 | |
XM_005254042.1 | 303 | Missense Mutation | AAG,GAG | K,E 62 | XP_005254099.1 |
DAOA-AS1 - DAOA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |