Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTTACACGAATCTAGAGGGCAC[A/G]GCCTACCACACACCTCGGCTGAACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609196 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372779 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105372779 - uncharacterized LOC105372779 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRAP - melanocortin 2 receptor accessory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285394.1 | Intron | NP_001272323.1 | ||||
NM_178817.3 | Intron | NP_848932.1 | ||||
NM_206898.1 | Intron | NP_996781.1 | ||||
XM_006724028.3 | Intron | XP_006724091.1 | ||||
XM_017028407.1 | Intron | XP_016883896.1 |