Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTCTAACGAGGAGCCTCTGCTG[C/T]TGAGCCAGAATCCTTTTCGGGTTCA
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 616023 MIM: 147450 | ||||||||||||||||||||||||||
Literature Links: |
SCAF4 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
SCAF4 - SR-related CTD associated factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145444.1 | 3805 | Missense Mutation | ACA,GCA | T,A 1127 | NP_001138916.1 | |
NM_001145445.1 | 3805 | Missense Mutation | ACA,GCA | T,A 1120 | NP_001138917.1 | |
NM_020706.2 | 3805 | Missense Mutation | ACA,GCA | T,A 1142 | NP_065757.1 | |
XM_005261017.2 | 3805 | Missense Mutation | ACA,GCA | T,A 1083 | XP_005261074.1 | |
XM_006724035.2 | 3805 | Missense Mutation | ACA,GCA | T,A 1138 | XP_006724098.1 | |
XM_006724036.2 | 3805 | Missense Mutation | ACA,GCA | T,A 1116 | XP_006724099.1 | |
XM_017028415.1 | 3805 | Missense Mutation | ACA,GCA | T,A 1141 | XP_016883904.1 | |
XM_017028416.1 | 3805 | Missense Mutation | ACA,GCA | T,A 1119 | XP_016883905.1 | |
XM_017028417.1 | 3805 | Missense Mutation | ACA,GCA | T,A 1061 | XP_016883906.1 |
SOD1 - superoxide dismutase 1, soluble | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |