Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCCTCTGATCGCCCTCTCAGC[C/T]CTTTCCTTCTGACTGAAAATCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DZANK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DZANK1 - double zinc ribbon and ankyrin repeat domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3192 - microRNA 3192 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR3F - RNA polymerase III subunit F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282526.1 | Intron | NP_001269455.1 | ||||
NM_006466.3 | Intron | NP_006457.2 | ||||
XM_011529141.1 | Intron | XP_011527443.1 | ||||
XM_017027596.1 | Intron | XP_016883085.1 |
Set Membership: |
HapMap |