Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCTCAGCCATCACCGAACTCAG[G/T]AAATATCTAAGGTATGTCTTTGTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600322 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNAP25 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SNAP25 - synaptosome associated protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322902.1 | 272 | Intron | NP_001309831.1 | |||
NM_001322903.1 | 272 | Intron | NP_001309832.1 | |||
NM_001322904.1 | 272 | Intron | NP_001309833.1 | |||
NM_001322905.1 | 272 | Intron | NP_001309834.1 | |||
NM_001322906.1 | 272 | Intron | NP_001309835.1 | |||
NM_001322907.1 | 272 | UTR 5 | NP_001309836.1 | |||
NM_001322908.1 | 272 | Intron | NP_001309837.1 | |||
NM_001322909.1 | 272 | Intron | NP_001309838.1 | |||
NM_001322910.1 | 272 | Intron | NP_001309839.1 | |||
NM_003081.4 | 272 | Intron | NP_003072.2 | |||
NM_130811.3 | 272 | Intron | NP_570824.1 | |||
XM_005260808.4 | 272 | Intron | XP_005260865.1 | |||
XM_017028021.1 | 272 | Intron | XP_016883510.1 | |||
XM_017028022.1 | 272 | Intron | XP_016883511.1 | |||
XM_017028023.1 | 272 | Intron | XP_016883512.1 |
SNAP25-AS1 - SNAP25 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |