Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCATGCTGTTTCCAGAGTTCCCC[A/G]TGGGATGGAGCTCCAGGTGCCTACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609935 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EBF4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EBF4 - early B-cell factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110514.1 | Intron | NP_001103984.1 | ||||
XM_006723600.1 | Intron | XP_006723663.1 | ||||
XM_017027983.1 | Intron | XP_016883472.1 | ||||
XM_017027984.1 | Intron | XP_016883473.1 | ||||
XM_017027985.1 | Intron | XP_016883474.1 |
Set Membership: |
HapMap |