Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTTAGTGTTCTTTTGTTTTTG[C/T]TTTGTTTATATTACAAACAATACTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611437 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DUSP19 - dual specificity phosphatase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142314.1 | Intron | NP_001135786.1 | ||||
NM_001321519.1 | Intron | NP_001308448.1 | ||||
NM_080876.3 | Intron | NP_543152.1 |
Set Membership: |
HapMap |