Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAGTCAGAGCTGTGGCACATAGA[C/T]TAAGCCAATCCACAAGTCAGTCAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608555 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTX2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MTX2 - metaxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006635.2 | Intron | NP_001006636.1 | ||||
NM_001319097.1 | Intron | NP_001306026.1 | ||||
NM_001319098.1 | Intron | NP_001306027.1 | ||||
NM_006554.4 | Intron | NP_006545.1 | ||||
XM_011510497.2 | Intron | XP_011508799.1 |