Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTACCTTTTTCTTTTTTAAACA[C/T]GTTTAATGTTGTACATGTACTATAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300395 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
THOC2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
THOC2 - THO complex 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081550.1 | 5651 | UTR 3 | NP_001075019.1 | |||
XM_005262450.3 | 5651 | Intron | XP_005262507.1 | |||
XM_017029662.1 | 5651 | UTR 3 | XP_016885151.1 | |||
XM_017029663.1 | 5651 | Intron | XP_016885152.1 | |||
XM_017029664.1 | 5651 | UTR 3 | XP_016885153.1 | |||
XM_017029665.1 | 5651 | Intron | XP_016885154.1 | |||
XM_017029666.1 | 5651 | UTR 3 | XP_016885155.1 | |||
XM_017029667.1 | 5651 | UTR 3 | XP_016885156.1 | |||
XM_017029668.1 | 5651 | Intron | XP_016885157.1 | |||
XM_017029669.1 | 5651 | UTR 3 | XP_016885158.1 | |||
XM_017029670.1 | 5651 | UTR 3 | XP_016885159.1 | |||
XM_017029671.1 | 5651 | UTR 3 | XP_016885160.1 | |||
XM_017029672.1 | 5651 | UTR 3 | XP_016885161.1 | |||
XM_017029673.1 | 5651 | UTR 3 | XP_016885162.1 | |||
XM_017029674.1 | 5651 | UTR 3 | XP_016885163.1 | |||
XM_017029675.1 | 5651 | UTR 3 | XP_016885164.1 | |||
XM_017029676.1 | 5651 | Intron | XP_016885165.1 | |||
XM_017029677.1 | 5651 | UTR 3 | XP_016885166.1 | |||
XM_017029678.1 | 5651 | UTR 3 | XP_016885167.1 | |||
XM_017029679.1 | 5651 | UTR 3 | XP_016885168.1 |
Set Membership: |
HapMap |