Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616512 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF152 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF152 - ring finger protein 152 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173557.2 | 6834 | Intron | NP_775828.1 | |||
XM_005266650.3 | 6834 | UTR 3 | XP_005266707.1 | |||
XM_005266652.3 | 6834 | UTR 3 | XP_005266709.1 | |||
XM_011525878.2 | 6834 | UTR 3 | XP_011524180.1 | |||
XM_011525879.2 | 6834 | UTR 3 | XP_011524181.1 | |||
XM_017025612.1 | 6834 | UTR 3 | XP_016881101.1 | |||
XM_017025613.1 | 6834 | UTR 3 | XP_016881102.1 |
Set Membership: |
HapMap |