Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCCTGTACGCCTCTGCTCTCTAC[C/T]GCCAGCGTCGTTCTGGCCGGGCCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100710 MIM: 601514 MIM: 613936 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHRNB1 - cholinergic receptor nicotinic beta 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FGF11 - fibroblast growth factor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303460.1 | 1081 | Missense Mutation | CGC,TGC | R,C 104 | NP_001290389.1 | |
NM_004112.3 | 1081 | Missense Mutation | CGC,TGC | R,C 163 | NP_004103.1 |
LOC107983988 - proteoglycan 4-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM102 - transmembrane protein 102 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |