Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600819 | ||||||||||||||||||||
Literature Links: |
FXR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FXR1 - FMR1 autosomal homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013438.2 | Intron | NP_001013456.1 | ||||
NM_001013439.2 | Intron | NP_001013457.1 | ||||
NM_005087.3 | Intron | NP_005078.2 | ||||
XM_005247813.3 | Intron | XP_005247870.2 | ||||
XM_005247814.3 | Intron | XP_005247871.2 | ||||
XM_005247815.3 | Intron | XP_005247872.2 | ||||
XM_005247816.3 | Intron | XP_005247873.2 | ||||
XM_006713775.3 | Intron | XP_006713838.1 | ||||
XM_017007298.1 | Intron | XP_016862787.1 | ||||
XM_017007299.1 | Intron | XP_016862788.1 | ||||
XM_017007300.1 | Intron | XP_016862789.1 | ||||
XM_017007301.1 | Intron | XP_016862790.1 |