Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114106 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP3CB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPP3CB - protein phosphatase 3 catalytic subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142353.2 | Intron | NP_001135825.1 | ||||
NM_001142354.2 | Intron | NP_001135826.1 | ||||
NM_001289968.1 | Intron | NP_001276897.1 | ||||
NM_001289969.1 | Intron | NP_001276898.1 | ||||
NM_021132.3 | Intron | NP_066955.1 | ||||
XM_005269944.1 | Intron | XP_005270001.1 | ||||
XM_017016379.1 | Intron | XP_016871868.1 |
Set Membership: |
HapMap |