Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601409 MIM: 610330 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAT5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KAT5 - lysine acetyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206833.1 | 1970 | Intron | NP_001193762.1 | |||
NM_006388.3 | 1970 | Intron | NP_006379.2 | |||
NM_182709.2 | 1970 | Intron | NP_874368.1 | |||
NM_182710.2 | 1970 | Intron | NP_874369.1 | |||
XM_006718421.2 | 1970 | Intron | XP_006718484.1 |
RNASEH2C - ribonuclease H2 subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032193.3 | 1970 | UTR 3 | NP_115569.2 |
Set Membership: |
HapMap |