Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGCCGGGTGGGGATGAGATGGGG[G/A]GCGCGGAGGATACAGGACACCCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300651 | ||||||||||||||||||||
Literature Links: |
LOC101927635 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927635 - uncharacterized LOC101927635 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PORCN - porcupine homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282167.1 | Intron | NP_001269096.1 | ||||
NM_022825.3 | Intron | NP_073736.2 | ||||
NM_203473.2 | Intron | NP_982299.1 | ||||
NM_203474.1 | Intron | NP_982300.1 | ||||
NM_203475.2 | Intron | NP_982301.1 | ||||
XM_005272635.2 | Intron | XP_005272692.1 | ||||
XM_005272636.2 | Intron | XP_005272693.1 | ||||
XM_005272637.1 | Intron | XP_005272694.1 | ||||
XM_006724545.3 | Intron | XP_006724608.1 | ||||
XM_006724546.3 | Intron | XP_006724609.1 | ||||
XM_006724547.1 | Intron | XP_006724610.1 | ||||
XM_011543948.2 | Intron | XP_011542250.1 | ||||
XM_017029735.1 | Intron | XP_016885224.1 | ||||
XM_017029736.1 | Intron | XP_016885225.1 |