Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603862 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCNT2 - cyclin T2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP3K19 - mitogen-activated protein kinase kinase kinase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018044.2 | 4882 | Intron | NP_001018054.1 | |||
NM_001018046.2 | 4882 | Intron | NP_001018056.1 | |||
NM_001018047.2 | 4882 | Intron | NP_001018057.1 | |||
NM_001282883.1 | 4882 | Intron | NP_001269812.1 | |||
NM_001321177.1 | 4882 | Intron | NP_001308106.1 | |||
NM_025052.4 | 4882 | Intron | NP_079328.3 | |||
XM_011511891.2 | 4882 | UTR 3 | XP_011510193.1 | |||
XM_011511892.1 | 4882 | Intron | XP_011510194.1 | |||
XM_011511894.2 | 4882 | UTR 3 | XP_011510196.1 | |||
XM_011511896.2 | 4882 | UTR 3 | XP_011510198.1 | |||
XM_011511897.2 | 4882 | UTR 3 | XP_011510199.1 | |||
XM_017005003.1 | 4882 | Intron | XP_016860492.1 | |||
XM_017005004.1 | 4882 | UTR 3 | XP_016860493.1 | |||
XM_017005005.1 | 4882 | UTR 3 | XP_016860494.1 | |||
XM_017005006.1 | 4882 | UTR 3 | XP_016860495.1 | |||
XM_017005007.1 | 4882 | Intron | XP_016860496.1 |