Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125305 MIM: 603725 MIM: 608073 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMTN PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DMTN - dematin actin binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FGF17 - fibroblast growth factor 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304478.1 | Intron | NP_001291407.1 | ||||
NM_003867.3 | Intron | NP_003858.1 | ||||
XM_005273675.1 | Intron | XP_005273732.1 | ||||
XM_011544683.1 | Intron | XP_011542985.1 | ||||
XM_011544684.2 | Intron | XP_011542986.1 | ||||
XM_011544685.1 | Intron | XP_011542987.1 |
NPM2 - nucleophosmin/nucleoplasmin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |