Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300579 | ||||||||||||||||||||
Literature Links: |
SHROOM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHROOM4 - shroom family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020717.3 | Intron | NP_065768.2 | ||||
XM_017029682.1 | Intron | XP_016885171.1 | ||||
XM_017029683.1 | Intron | XP_016885172.1 | ||||
XM_017029684.1 | Intron | XP_016885173.1 | ||||
XM_017029685.1 | Intron | XP_016885174.1 | ||||
XM_017029686.1 | Intron | XP_016885175.1 | ||||
XM_017029687.1 | Intron | XP_016885176.1 |