Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKUB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKUB1 - ankyrin repeat and ubiquitin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144960.2 | 1106 | Missense Mutation | ATG,ATT | M,I 306 | NP_001138432.1 | |
NM_001315505.1 | 1106 | Missense Mutation | ATG,ATT | M,I 160 | NP_001302434.1 | |
NM_001315506.1 | 1106 | Missense Mutation | ATG,ATT | M,I 306 | NP_001302435.1 | |
XM_011512798.1 | 1106 | Missense Mutation | ATG,ATT | M,I 257 | XP_011511100.1 | |
XM_011512799.1 | 1106 | Missense Mutation | ATG,ATT | M,I 306 | XP_011511101.1 | |
XM_011512801.1 | 1106 | Missense Mutation | ATG,ATT | M,I 140 | XP_011511103.1 | |
XM_011512802.1 | 1106 | Intron | XP_011511104.1 |