Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608614 MIM: 229000 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4V2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP4V2 - cytochrome P450 family 4 subfamily V member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLKB1 - kallikrein B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000892.4 | Intron | NP_000883.2 | ||||
NM_001318394.1 | Intron | NP_001305323.1 | ||||
NM_001318396.1 | Intron | NP_001305325.1 | ||||
XM_011531930.2 | Intron | XP_011530232.1 | ||||
XM_017008181.1 | Intron | XP_016863670.1 | ||||
XM_017008182.1 | Intron | XP_016863671.1 | ||||
XM_017008183.1 | Intron | XP_016863672.1 | ||||
XM_017008184.1 | Intron | XP_016863673.1 |
Set Membership: |
HapMap |