Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604933 MIM: 604746 MIM: 613931 | ||||||||||||||||||||
Literature Links: |
MUTYH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TESK2 - testis-specific kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOE1 - target of EGR1, member 1 (nuclear) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025077.3 | 284 | Intron | NP_079353.3 | |||
XM_005270412.3 | 284 | Intron | XP_005270469.1 | |||
XM_005270413.4 | 284 | Intron | XP_005270470.1 | |||
XM_011540569.2 | 284 | Intron | XP_011538871.1 |