Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCCCCCGTGTCACCGTCCTGGTG[C/T]GGGAGTTCGAGGCATTTGACAACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606596 MIM: 109190 MIM: 614767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKRP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKRP - fukutin related protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039885.2 | 306 | Missense Mutation | CGG,TGG | R,W 54 | NP_001034974.1 | |
NM_024301.4 | 306 | Missense Mutation | CGG,TGG | R,W 54 | NP_077277.1 | |
XM_005259247.2 | 306 | Missense Mutation | CGG,TGG | R,W 54 | XP_005259304.1 | |
XM_005259248.2 | 306 | Missense Mutation | CGG,TGG | R,W 54 | XP_005259305.1 | |
XM_005259249.4 | 306 | Missense Mutation | CGG,TGG | R,W 54 | XP_005259306.1 | |
XM_011527306.1 | 306 | Missense Mutation | CGG,TGG | R,W 54 | XP_011525608.1 | |
XM_011527307.1 | 306 | Missense Mutation | CGG,TGG | R,W 54 | XP_011525609.1 | |
XM_017027297.1 | 306 | Missense Mutation | CGG,TGG | R,W 54 | XP_016882786.1 |
SLC1A5 - solute carrier family 1 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRN4 - striatin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |