Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608023 MIM: 612038 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX56 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDX56 - DEAD-box helicase 56 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMED4 - transmembrane p24 trafficking protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303058.1 | 1181 | Intron | NP_001289987.1 | |||
NM_001303059.1 | 1181 | UTR 3 | NP_001289988.1 | |||
NM_001303060.1 | 1181 | Intron | NP_001289989.1 | |||
NM_001303061.1 | 1181 | UTR 3 | NP_001289990.1 | |||
NM_001303062.1 | 1181 | UTR 3 | NP_001289991.1 | |||
NM_182547.3 | 1181 | UTR 3 | NP_872353.2 |
Set Membership: |
HapMap |