Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCAF1 - DDB1 and CUL4 associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171904.1 | Intron | NP_001165375.1 | ||||
NM_014703.2 | Intron | NP_055518.1 | ||||
XM_005276753.4 | Intron | XP_005276810.1 | ||||
XM_005276755.4 | Intron | XP_005276812.1 | ||||
XM_011534273.2 | Intron | XP_011532575.1 | ||||
XM_011534274.2 | Intron | XP_011532576.1 | ||||
XM_011534275.2 | Intron | XP_011532577.1 | ||||
XM_011534276.2 | Intron | XP_011532578.1 | ||||
XM_011534277.2 | Intron | XP_011532579.1 | ||||
XM_017007546.1 | Intron | XP_016863035.1 | ||||
XM_017007547.1 | Intron | XP_016863036.1 | ||||
XM_017007548.1 | Intron | XP_016863037.1 | ||||
XM_017007549.1 | Intron | XP_016863038.1 | ||||
XM_017007550.1 | Intron | XP_016863039.1 | ||||
XM_017007551.1 | Intron | XP_016863040.1 |