Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605719 MIM: 600404 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LAT2 - linker for activation of T-cells family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFC2 - replication factor C subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278791.1 | Intron | NP_001265720.1 | ||||
NM_001278792.1 | Intron | NP_001265721.1 | ||||
NM_001278793.1 | Intron | NP_001265722.1 | ||||
NM_002914.4 | Intron | NP_002905.2 | ||||
NM_181471.2 | Intron | NP_852136.1 | ||||
XM_006716080.2 | Intron | XP_006716143.1 | ||||
XM_017012492.1 | Intron | XP_016867981.1 | ||||
XM_017012493.1 | Intron | XP_016867982.1 |
Set Membership: |
HapMap |