Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602704 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MDM4 - MDM4, p53 regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204171.1 | 66 | UTR 5 | NP_001191100.1 | |||
NM_001204172.1 | 66 | UTR 5 | NP_001191101.1 | |||
NM_001278516.1 | 66 | UTR 5 | NP_001265445.1 | |||
NM_001278517.1 | 66 | UTR 5 | NP_001265446.1 | |||
NM_001278518.1 | 66 | UTR 5 | NP_001265447.1 | |||
NM_001278519.1 | 66 | UTR 5 | NP_001265448.1 | |||
NM_002393.4 | 66 | UTR 5 | NP_002384.2 | |||
XM_017001311.1 | 66 | Intron | XP_016856800.1 | |||
XM_017001312.1 | 66 | Intron | XP_016856801.1 | |||
XM_017001313.1 | 66 | Intron | XP_016856802.1 |