Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATTGTCTTCTTCCAGCCAGTTT[C/G]TAAGGCAGGCAATGGAAACAGGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613299 MIM: 613300 | ||||||||||||||||||||
Literature Links: |
FAM13A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM13A - family with sequence similarity 13 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015045.2 | 2531 | UTR 3 | NP_001015045.1 | |||
NM_001265578.1 | 2531 | UTR 3 | NP_001252507.1 | |||
NM_001265579.1 | 2531 | UTR 3 | NP_001252508.1 | |||
NM_001265580.1 | 2531 | UTR 3 | NP_001252509.1 | |||
NM_014883.3 | 2531 | UTR 3 | NP_055698.2 |
FAM13A-AS1 - FAM13A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |