Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607968 | ||||||||||||||||||||
Literature Links: |
BBS9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BBS9 - Bardet-Biedl syndrome 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033604.1 | Intron | NP_001028776.1 | ||||
NM_001033605.1 | Intron | NP_001028777.1 | ||||
NM_014451.3 | Intron | NP_055266.2 | ||||
NM_198428.2 | Intron | NP_940820.1 | ||||
XM_005249700.3 | Intron | XP_005249757.1 | ||||
XM_005249701.2 | Intron | XP_005249758.1 | ||||
XM_011515264.2 | Intron | XP_011513566.1 | ||||
XM_011515265.2 | Intron | XP_011513567.1 | ||||
XM_011515266.2 | Intron | XP_011513568.1 | ||||
XM_011515267.2 | Intron | XP_011513569.1 | ||||
XM_011515268.1 | Intron | XP_011513570.1 | ||||
XM_011515269.2 | Intron | XP_011513571.1 | ||||
XM_011515270.2 | Intron | XP_011513572.1 | ||||
XM_017011990.1 | Intron | XP_016867479.1 | ||||
XM_017011991.1 | Intron | XP_016867480.1 | ||||
XM_017011992.1 | Intron | XP_016867481.1 | ||||
XM_017011993.1 | Intron | XP_016867482.1 | ||||
XM_017011994.1 | Intron | XP_016867483.1 |