Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 143110 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLA-F PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HLA-F - major histocompatibility complex, class I, F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098478.1 | 447 | Silent Mutation | TAA,TAC | *,Y 139 | NP_001091948.1 | |
NM_001098479.1 | 447 | Silent Mutation | TAA,TAC | *,Y 139 | NP_001091949.1 | |
NM_018950.2 | 447 | Silent Mutation | TAA,TAC | *,Y 139 | NP_061823.2 | |
XM_011514564.1 | 447 | Nonsense Mutation | TAA,TAC | *,Y 139 | XP_011512866.1 | |
XM_017010810.1 | 447 | Nonsense Mutation | TAA,TAC | *,Y 139 | XP_016866299.1 | |
XM_017010811.1 | 447 | Nonsense Mutation | TAA,TAC | *,Y 139 | XP_016866300.1 | |
XM_017010812.1 | 447 | Nonsense Mutation | TAA,TAC | *,Y 139 | XP_016866301.1 | |
XM_017010813.1 | 447 | Nonsense Mutation | TAA,TAC | *,Y 139 | XP_016866302.1 | |
XM_017010814.1 | 447 | Silent Mutation | TAA,TAC | *,Y 21 | XP_016866303.1 | |
XM_017010815.1 | 447 | Nonsense Mutation | TAA,TAC | *,Y 21 | XP_016866304.1 |
HLA-F-AS1 - HLA-F antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |