Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116899 | ||||||||||||||||||||
Literature Links: |
CDKN1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDKN1A - cyclin dependent kinase inhibitor 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000389.4 | Intron | NP_000380.1 | ||||
NM_001220777.1 | Intron | NP_001207706.1 | ||||
NM_001220778.1 | Intron | NP_001207707.1 | ||||
NM_001291549.1 | Intron | NP_001278478.1 | ||||
NM_078467.2 | Intron | NP_510867.1 |
PANDAR - promoter of CDKN1A antisense DNA damage activated RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |