Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK1 - cyclin dependent kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170406.1 | Intron | NP_001163877.1 | ||||
NM_001170407.1 | Intron | NP_001163878.1 | ||||
NM_001320918.1 | Intron | NP_001307847.1 | ||||
NM_001786.4 | Intron | NP_001777.1 | ||||
NM_033379.4 | Intron | NP_203698.1 | ||||
XM_005270303.3 | Intron | XP_005270360.1 |