Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116940 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDK1 - cyclin dependent kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170406.1 | Intron | NP_001163877.1 | ||||
NM_001170407.1 | Intron | NP_001163878.1 | ||||
NM_001320918.1 | Intron | NP_001307847.1 | ||||
NM_001786.4 | Intron | NP_001777.1 | ||||
NM_033379.4 | Intron | NP_203698.1 | ||||
XM_005270303.3 | Intron | XP_005270360.1 |
Set Membership: |
HapMap |