Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606015 MIM: 604136 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCMR PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FCMR - Fc fragment of IgM receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL24 - interleukin 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185156.1 | Intron | NP_001172085.1 | ||||
NM_001185157.1 | Intron | NP_001172086.1 | ||||
NM_001185158.1 | Intron | NP_001172087.1 | ||||
NM_006850.3 | Intron | NP_006841.1 | ||||
XM_011509101.2 | Intron | XP_011507403.1 | ||||
XM_017000121.1 | Intron | XP_016855610.1 |