Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCCATGGGCTATGAGCGAGAGC[A/G]GGTCGTGGCCGCCCTGAGAGCCAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109091 MIM: 605162 MIM: 600061 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALR PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CALR - calreticulin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GADD45GIP1 - GADD45G interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6515 - microRNA 6515 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAD23A - RAD23 homolog A, nucleotide excision repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270362.1 | 671 | Missense Mutation | CAG,CGG | Q,R 179 | NP_001257291.1 | |
NM_001270363.1 | 671 | Missense Mutation | CAG,CGG | Q,R 179 | NP_001257292.1 | |
NM_005053.3 | 671 | Missense Mutation | CAG,CGG | Q,R 179 | NP_005044.1 |
Set Membership: |
HapMap |