Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300117 | ||||||||||||||||||||
Literature Links: |
FAM133A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM133A - family with sequence similarity 133 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171109.1 | Intron | NP_001164580.1 | ||||
NM_001171110.1 | Intron | NP_001164581.1 | ||||
NM_001171111.1 | Intron | NP_001164582.1 | ||||
NM_173698.2 | Intron | NP_775969.1 | ||||
XM_006724641.3 | Intron | XP_006724704.1 | ||||
XM_011530925.2 | Intron | XP_011529227.1 | ||||
XM_011530926.2 | Intron | XP_011529228.1 | ||||
XM_011530927.2 | Intron | XP_011529229.1 | ||||
XM_011530928.1 | Intron | XP_011529230.1 | ||||
XM_011530929.1 | Intron | XP_011529231.1 | ||||
XM_011530931.2 | Intron | XP_011529233.1 | ||||
XM_017029454.1 | Intron | XP_016884943.1 | ||||
XM_017029455.1 | Intron | XP_016884944.1 | ||||
XM_017029456.1 | Intron | XP_016884945.1 |
NAP1L3 - nucleosome assembly protein 1 like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |