Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186590 MIM: 615733 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJC30 - DnaJ heat shock protein family (Hsp40) member C30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX1A - syntaxin 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBSCR22 - Williams-Beuren syndrome chromosome region 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202560.2 | Intron | NP_001189489.1 | ||||
NM_017528.4 | Intron | NP_059998.2 | ||||
XM_006715847.1 | Intron | XP_006715910.1 | ||||
XM_011515778.1 | Intron | XP_011514080.1 | ||||
XM_011515779.1 | Intron | XP_011514081.1 |
Set Membership: |
HapMap |